3-Bromo-6-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-6-nitroindazole

OR14085-1G OR14085-5G
76566-702EA 26 USD
76566-702 76566-704
3-Bromo-6-nitro-1H-indazole
3-Bromo-6-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD06410881
CAS Number: 70315-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR