N-Benzyl-2-cyanoacetamide

Supplier: APOLLO SCIENTIFIC

Warning

OR14083-1G OR14083-5G
76566-700EA 110 USD
76566-700 76566-698
N-Benzyl-2-cyanoacetamide
N-Benzyl-2-cyanoacetamide
Formula: C₁₀H₁₀N₂O
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00461336
CAS Number: 10412-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR