4-Bromobenzo[b]thiophene-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC

OR14031-1G
76566-656EA 48 USD
76566-656
4-Bromobenzo[b]thiophene-2-carboxylic acid
4-Bromobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₅BrO₂S
MW: 257.1038 g/mol
Melting Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD03407342
CAS Number: 5194-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR