3-(1H-Pyrazol-3-yl)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

OR1400-250MG OR1400-1G
76566-652EA 37 USD
76566-652 76566-654
3-(1H-Pyrazol-3-yl)benzoic acid 97%
3-(1H-Pyrazol-3-yl)benzoic acid
Formula: C₁₀H₈N₂O₂
Melting Pt: 203…204 °C
Storage Temperature: Ambient
MDL Number: MFCD03407956
CAS Number: 850375-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR