4'-Hydroxybiphenyl-4-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4'-Hydroxy-4-biphenylcarboxylic acid4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

OR13999-100G OR13999-5G OR13999-25G
76566-646EA 18 USD
76566-646 76566-648 76566-650
4'-Hydroxybiphenyl-4-carboxylic acid 95%
4'-Hydroxybiphenyl-4-carboxylic acid
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00059078
CAS Number: 58574-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR