2-Amino-4-methoxypyridine

Supplier: APOLLO SCIENTIFIC

Warning

OR13856-5G OR13856-25G
76566-594EA 33 USD
76566-594 76566-596
2-Amino-4-methoxypyridine
2-Amino-4-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07437849
CAS Number: 10201-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR