Methyl-5-bromo-6-chloronicotinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 5-bromo-6-chloronicotinate

OR13854-1G OR13854-25G OR13854-5G
76566-590EA 30 USD
76566-590 76566-592 76566-588
Methyl-5-bromo-6-chloronicotinate
Methyl 5-bromo-6-chloronicotinate
Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844855
CAS Number: 78686-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR