3-Bromo-1H-1,2,4-triazole

Supplier: APOLLO SCIENTIFIC

Warning

OR13675-25G OR13675-5G
76566-492EA 21 USD
76566-492 76566-494
3-Bromo-1H-1,2,4-triazole
3-Bromo-1H-1,2,4-triazole
Formula: C₂H₂BrN₃
MW: 147.9614 g/mol
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD01421960
CAS Number: 7343-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR