5-Bromo-7-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR13583-250MG OR13583-1G OR13583-5G
76566-406EA 13 USD
76566-406 76566-408 76566-410
5-Bromo-7-methyl-1H-indazole
5-Bromo-7-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD03990484
CAS Number: 156454-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR