2,4,6-Trimethylbenzenesulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR1357-5G OR1357-25G
76566-400EA 13 USD
76566-400 76566-402
2,4,6-Trimethylbenzenesulfonamide
2,4,6-Trimethylbenzenesulphonamide
Formula: C₉H₁₃NO₂S
MW: 199.2700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463245
CAS Number: 4543-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR