2-Quinolinecarbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Quinoline-2-carbonitrile

OR13512-5G OR13512-25G OR13512-1G
76566-380EA 70 USD
76566-380 76566-382 76566-378
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile
Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Boiling Pt: 160 °C/23mmHg
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00134341
CAS Number: 1436-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR