3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

OR13238-1G OR13238-250MG
76566-264EA 22 USD
76566-264 76566-266
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde
Formula: C₉H₉NO₅
MW: 211.1715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058678
CAS Number: 178686-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR