2-Amino-5-bromopyrazine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5-bromopyrazin-2-amine

OR13181-10G OR13181-5G OR13181-25G
76566-198EA 15 USD
76566-198 76566-200 76566-202
2-Amino-5-bromopyrazine
2-Amino-5-bromopyrazine
Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00235015
CAS Number: 59489-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR