4-Chloro-3-nitrophenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Chloro-3-nitrobenzeneboronic acid

OR13107-1G OR13107-25G OR13107-5G
76566-132EA 20 USD
76566-132 76566-134 76566-136
4-Chloro-3-nitrophenylboronic acid
4-Chloro-3-nitrophenylboronic acid
Formula: C₆H₅BClNO₄
MW: 201.37 g/mol
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD02258950
CAS Number: 151169-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR