Methyl-5-bromopicolinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 5-bromopyridine-2-carboxylateMethyl 5-bromopicolinate

OR13058-5G OR13058-1G OR13058-25G
76566-090EA 13 USD
76566-090 76566-092 76566-094
Methyl-5-bromopicolinate
Methyl 5-bromopicolinate
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD04112493
CAS Number: 29682-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR