4-Hydroxy-3-methoxybenzylamine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Aminomethyl)-2-methoxyphenol hydrochloride

OR13027-10G OR13027-1G
76560-212EA 14.35 USD
76560-212 76560-472
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00012864
CAS Number: 7149-10-2
EINECS: 230-468-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR