1-Methyl-1H-pyrazol-4-carbaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Methyl-1H-pyrazole-4-carbaldehyde1-Methyl-1H-pyrazole-4-carboxaldehyde

OR12622-10G OR12622-1G OR12622-5G
76565-836EA 66 USD
76565-836 76565-834 76565-832
1-Methyl-1H-pyrazol-4-carbaldehyde
1-Methyl-1H-pyrazol-4-carbaldehyde
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 29…34 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00460465
CAS Number: 25016-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR