4-Phenoxypiperidine hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

OR12597-1G OR12597-250MG
76565-812EA 21 USD
76565-812 76565-810
4-Phenoxypiperidine hydrochloride
4-Phenoxypiperidine hydrochloride
Formula: C₁₁H₁₆ClNO
MW: 213.7038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684106
CAS Number: 3413-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR