1-Isoquinolinecarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Isoquinoline-1-carboxylic acid

OR12594-250MG OR12594-25G OR12594-5G
76565-806EA 48 USD
76565-806 76565-804 76565-808
1-Isoquinolinecarboxylic acid 98%
1-Isoquinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006901
CAS Number: 486-73-7
EINECS: 207-639-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR