5-(4-Chlorophenyl)oxazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-(4-Chlorophenyl)-1,3-oxazole

OR12571-5G OR12571-1G
76565-798EA 313 USD
76565-798 76565-796
5-(4-Chlorophenyl)oxazole
5-(4-Chlorophenyl)oxazole
Formula: C₉H₆ClNO
MW: 179.6030 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258870
CAS Number: 1008-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR