3-(2-Pyrrolidinyl)pyridine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: DL-Nornicotine2-(3-Pyridyl)pyrrolidine(±)-Nornicotine

OR12516-250MG OR12516-1G
76565-772EA 19 USD
76565-772 76565-774
3-(2-Pyrrolidinyl)pyridine
3-(2-Pyrrolidinyl)pyridine
Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Boiling Pt: 107…108 °C/2mmHg
Density: 1.074 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016913
CAS Number: 5746-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR