(2-Chloro-4-pyridinyl)methanol 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (2-Chloropyridin-4-yl)methanol2-Chloropyridine-4-methanol

OR12246-5G
76565-690EA 61 USD
76565-690
(2-Chloro-4-pyridinyl)methanol 97%
(2-Chloro-4-pyridinyl)methanol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD06858767
CAS Number: 100704-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR