4-Bromo-1-methylimidazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1-methyl-1H-imidazole

OR1221-1G OR1221-10G OR1221-5G
76565-680EA 169 USD
76565-680 76565-682 76565-678
4-Bromo-1-methylimidazole 98%
4-Bromo-1-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
Boiling Pt: 91…93 °C/0.1mmHg
Density: 1.614 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01320501
CAS Number: 25676-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR