Methyl-4-(3-bromopropoxy)benzenecarboxylate

Supplier: APOLLO SCIENTIFIC

Warning

OR12129-5G OR12129-1G
76565-634EA 257 USD
76565-634 76565-632
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Formula: C₁₁H₁₃BrO₃
MW: 273.13 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD01814575
CAS Number: 135998-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR