Methyl-2-aminothiazole-4-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-aminothiazole-4-carboxylate

OR1210-100G OR1210-25G
76565-614EA 58 USD
76565-614 76565-612
Methyl-2-aminothiazole-4-carboxylate 98%
Methyl 2-aminothiazole-4-carboxylate
Formula: C₅H₆N₂O₂S
MW: 158.1800 g/mol
Melting Pt: 171…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00622441
CAS Number: 118452-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR