3-Bromo-5-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-5-nitroindazole

OR12046-5G
76565-560EA 18 USD
76565-560
3-Bromo-5-nitro-1H-indazole
3-Bromo-5-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Melting Pt: 227…229 °C
Storage Temperature: Ambient
MDL Number: MFCD07367964
CAS Number: 67400-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR