1-Bromo-4,5-dichloro-2-nitrobenzene

Supplier: APOLLO SCIENTIFIC

Warning

OR11999-25G OR11999-1G OR11999-5G
76562-190EA 53 USD
76562-190 76562-826 76560-102
1-Bromo-4,5-dichloro-2-nitrobenzene
1-Bromo-4,5-dichloro-2-nitrobenzene
Formula: C₆H₂BrCl₂NO₂
MW: 270.9000 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD07780670
CAS Number: 93361-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR