2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone

Supplier: APOLLO SCIENTIFIC

Warning

OR11952-1G OR11952-250MG
76565-490EA 147 USD
76565-490 76565-488
2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone
2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone
Formula: C₁₂H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD09800919
CAS Number: 951884-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR