2-Chloro-5-nitronicotinic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR11941-250MG
76565-478EA 13 USD
76565-478
2-Chloro-5-nitronicotinic acid 95%
2-Chloro-5-nitronicotinic acid
Formula: C₆H₃ClN₂O₄
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD06658402
CAS Number: 42959-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR