5-Methyl-2-(2-nitrophenyl)oxazole

Supplier: APOLLO SCIENTIFIC

Warning

OR11929-500MG OR11929-100MG OR11929-1G
76565-454EA 74 USD
76565-454 76565-458 76565-456
5-Methyl-2-(2-nitrophenyl)oxazole
5-Methyl-2-(2-nitrophenyl)oxazole
Formula: C₁₀H₈N₂O₃
MW: 204.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801026
CAS Number: 52829-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR