4-Boronobenzenesulfonamide 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Sulphamoylbenzeneboronic acid(4-Aminosulfonylphenyl)boronic acid(4-Aminosulphonylphenyl)boronic acid4-Sulfamoylbenzeneboronic acid

OR1186-5G OR1186-1G
76565-438EA 252 USD
76565-438 76565-436
4-Boronobenzenesulfonamide 95%
4-Boronobenzenesulphonamide
Formula: C₆H₈BNO₄S
MW: 201.00 g/mol
Melting Pt: 250…256 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659839
CAS Number: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR