5-(3-Nitrophenyl)-4-oxazolecarboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5-(3-Nitrophenyl)-1,3-oxazole-4-carboxylic acid

OR11681-100MG OR11681-1G OR11681-500MG
76565-372EA 153 USD
76565-372 76565-370 76565-374
5-(3-Nitrophenyl)-4-oxazolecarboxylic acid
5-(3-Nitrophenyl)-4-oxazolecarboxylic acid
Formula: C₁₀H₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD09800992
CAS Number: 951885-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR