8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

OR11664-1G
76565-364EA 29 USD
76565-364
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
Formula: C₇H₅BrClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09800973
CAS Number: 957120-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR