8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

OR11663-250MG OR11663-1G
76565-362EA 147 USD
76565-362 76565-360
8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
Formula: C₈H₈BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD09800972
CAS Number: 957120-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR