2-Amino-6-chloropyrazine 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR11609-5G OR11609-100G OR11609-25G
76565-332EA 13 USD
76565-332 76565-336 76565-334
2-Amino-6-chloropyrazine 98%
2-Amino-6-chloropyrazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00055024
CAS Number: 33332-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR