3-Chloro-(5-methoxycarbonyl)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (3-Chloro-5-(methoxycarbonyl)phenyl)boronic acid

OR11535-100MG
76565-286EA 31 USD
76565-286
3-Chloro-(5-methoxycarbonyl)phenylboronic acid 97%
3-Chloro-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214.4107 g/mol
Melting Pt: 217…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09475906
CAS Number: 957120-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR