2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(3-(Dimethylamino)-1-methoxyallylidene)malononitrile

OR11440-1G
76563-400EA 13.33 USD
76563-400
2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile 98%
2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile
Formula: C₉H₁₁N₃O
MW: 177.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00975440
CAS Number: 95689-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR