4-(4-Methoxyphenoxy)aniline 98+%

Supplier: APOLLO SCIENTIFIC

Danger

OR11346-5G OR11346-1G
76565-146EA 47 USD
76565-146 76565-144
4-(4-Methoxyphenoxy)aniline 98+%
4-(4-Methoxyphenoxy)aniline
Formula: C₁₃H₁₃NO₂
MW: 215.2478 g/mol
Boiling Pt: 245…250 °C/33mmHg
Melting Pt: 75…79 °C
Density: 1.294 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044105
EINECS: 250-644-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR