5-Bromo-2'-chloro-2-hydroxybenzophenone

Supplier: APOLLO SCIENTIFIC

Warning

OR1128-10G OR1128-1G OR1128-5G
76565-060EA 427 USD
76565-060 76565-062 76565-058
5-Bromo-2'-chloro-2-hydroxybenzophenone
5-Bromo-2'-chloro-2-hydroxybenzophenone
Formula: C₁₃H₈BrClO₂
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD02682999
CAS Number: 332104-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR