5-Nitroindan

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Nitroindane

OR11226-5G OR11226-25G
76565-008EA 94 USD
76565-008 76565-006
5-Nitroindan
5-Nitroindan
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Storage Temperature: Ambient
CAS Number: 7436-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR