2,5-Dihydroxy-p-benzoquinone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,5-Dihydroxy-1,4-benzoquinone

OR11157-25G OR11157-100G OR11157-5G
76564-934EA 183 USD
76564-934 76564-930 76564-932
2,5-Dihydroxy-p-benzoquinone
2,5-Dihydroxy-p-benzoquinone
Formula: C₆H₄O₄
MW: 140.1 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00001598
CAS Number: 615-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR