3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR111401-250MG
76564-824EA 21 USD
76564-824
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid
Formula: C₈H₅BrN₂O₂
MW: 241.05 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD07021511
CAS Number: 886362-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR