3'-Bromo-5'-chloro-2'-hydroxyacetophenone

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-(3-Bromo-5-chloro-2-hydroxyphenyl)ethanone5-Chloro-3-bromo-2-hydroxyacetophenone

OR1112-25G OR1112-5G
76564-732EA 29 USD
76564-732 76564-734
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
Formula: C₈H₆BrClO₂
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00857068
CAS Number: 59443-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR