Luminol (3-Aminophthalhydrazide) 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-Amino-2,3-dihydrophthalazine-1,4-dione
, NSC 5064
, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione
OR11118-25G
76564-712EA
43
USD
76564-712
Luminol (3-Aminophthalhydrazide) 98%
Luminol (3-Aminophthalhydrazide)
Formula:
C₈H₇N₃O₂ MW: 177.16 g/mol Melting Pt: 319…320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006890 CAS Number: 521-31-3 EINECS: 208-309-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...