Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

OR110968-1G OR110968-250MG OR110968-5G
76564-580EA 104 USD
76564-580 76564-582 76564-584
Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₉H₇ClN₂O₂
MW: 210.62
Storage Temperature: Ambient
MDL Number: MFCD07021508
CAS Number: 900019-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR