3,5-Dibromo-4-methylbenzoic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR11090-5G OR11090-25G
76564-500EA 208 USD
76564-500 76564-498
3,5-Dibromo-4-methylbenzoic acid
3,5-Dibromo-4-methylbenzoic acid
Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017544
CAS Number: 67973-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR