Imidazo[1,2-a]pyridine-8-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR110823-1G
76564-402EA 24 USD
76564-402
Imidazo[1,2-a]pyridine-8-carboxylic acid
Imidazo[1,2-a]pyridine-8-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739251
CAS Number: 133427-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR