2-(4-Aminophenyl)propanoic acid

Supplier: APOLLO SCIENTIFIC

OR110788-1G
76564-382EA 157 USD
76564-382
2-(4-Aminophenyl)propanoic acid
2-(4-Aminophenyl)propanoic acid
Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00800233
CAS Number: 59430-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR