7-Nitro-2H-1,4-benzoxazin-3(4H)-one

Supplier: APOLLO SCIENTIFIC

Warning

OR110745-25G OR110745-1G OR110745-5G
76564-346EA 352 USD
76564-346 76564-342 76564-344
7-Nitro-2H-1,4-benzoxazin-3(4H)-one
7-Nitro-2H-1,4-benzoxazin-3(4H)-one
Formula: C₈H₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00090355
CAS Number: 81721-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR