Methyl-4-(hydroxymethyl)benzoate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 4-(hydroxymethyl)benzoate

OR110419-10G OR110419-25G OR110419-5G
76564-116EA 37 USD
76564-116 76564-118 76564-114
Methyl-4-(hydroxymethyl)benzoate
Methyl 4-(hydroxymethyl)benzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00009984
CAS Number: 6908-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR